This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01229841_g1
See other MST1 GE Assays ›
Gene Symbol:
Mst1
Gene Name:
macrophage stimulating 1 (hepatocyte growth factor-like)
Gene Aliases:
D3F15S2h, D9H3F15S2, DNF15S2h, Hgfl
Chromosome Location:
Chr.9: 108080380 - 108085027 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01229841_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mst1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_008243.3 NP_032269.3 8 - 9 1040 66
GenBank mRNA AK040934.1 - 8 - 9 1024 66
AK134222.1 - 8 - 9 1023 66
AK144824.1 - 8 - 9 1003 66
BC010551.1 - 8 - 9 1025 4195560 66
BC051393.1 - 8 - 9 1009 5055055 66
M74181.1 - 8 - 9 997 66

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0900001393.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Mst1

Panther Classification:

Molecular Function -

serine protease

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
embryo implantation
serine-type endopeptidase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0900011180.mm.1 NM_008243.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900011180.mm.1 NM_008243.3 Refseq
Affymetrix Mouse Clariom S Array PSR0900011180.mm.1 TC0900001393.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0