This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01229822_g1
See other GMPPB GE Assays ›
Gene Symbol:
Gmppb
Gene Name:
GDP-mannose pyrophosphorylase B
Gene Aliases:
AI317178, E430010H19
Chromosome Location:
Chr.9: 108049229 - 108051936 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01229822_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gmppb
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_177910.3 NP_808578.1 2 - 3 438 72
GenBank mRNA AK088295.1 - 2 - 3 420 72
AK148125.1 - 438 72
AK158470.1 - 2 - 3 438 72
AK165816.1 - 2 - 3 440 72
BC061207.1 - 3 - 4 312 30249330 72

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0900001391.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gmppb

Panther Classification:

Molecular Function -

transferase

Gene Ontology Categories:

Function(s) Process(es)

biosynthetic process
nucleotide binding
mannose-1-phosphate guanylyltransferase activity
GTP binding
transferase activity
nucleotidyltransferase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0900011147.mm.1 NM_177910.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900011147.mm.1 NM_177910.3 Refseq
Affymetrix Mouse Clariom S Array PSR0900011147.mm.1 TC0900001391.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0