This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01229754_g1
See other ACTR1B GE Assays ›
Gene Symbol:
Actr1b
Gene Name:
ARP1 actin-related protein 1B, centractin beta
Gene Aliases:
2310066K23Rik, AA960180, AI851923, Arp1b
Chromosome Location:
Chr.1: 36698114 - 36710017 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01229754_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Actr1b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_146107.2 NP_666219.1 7 - 8 823 150
GenBank mRNA AK078508.1 - 7 - 8 839 150
AK088839.1 - 7 - 8 856 150
AK178242.1 - 1 - 2 109 150
AK205839.1 - 2 - 3 190 150
BC023020.1 - 7 - 8 823 5375425 150
BC024861.1 - 2 - 3 147 150

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0100002189.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Actr1b

Panther Classification:

Molecular Function -

actin and actin related protein actin or actin-binding cytoskeletal protein cytoskeletal protein

Gene Ontology Categories:

Function(s) Process(es)

biological_process
nucleotide binding
molecular_function
ATP binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100017574.mm.1 NM_146107.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100017574.mm.1 NM_146107.2 Refseq
Affymetrix Mouse Clariom S Array PSR0100017574.mm.1 TC0100002189.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100017573.mm.1 NM_146107.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100017573.mm.1 NM_146107.2 Refseq
Affymetrix Mouse Clariom S Array PSR0100017573.mm.1 TC0100002189.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0