This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01229747_m1
See other 4933424G06RIK GE Assays ›
Gene Symbol:
4933424G06Rik
Gene Name:
RIKEN cDNA 4933424G06 gene
Gene Aliases:
1700104I15Rik
Chromosome Location:
Chr.1: 36714538 - 36757960 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01229747_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 4933424G06Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_040290.1 6 - 7 601 62
GenBank mRNA AK016893.1 - 6 - 7 601 62

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0100000254.mm.1

Set Membership:

Amplicon length less than or equal to 70 Mus musculus Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100001705.mm.1 NR_040290.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100001705.mm.1 NR_040290.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100001705.mm.1 TC0100000254.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100001709.mm.1 NR_040290.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100001709.mm.1 NR_040290.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100001709.mm.1 TC0100000254.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100001711.mm.1 NR_040290.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100001711.mm.1 NR_040290.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100001711.mm.1 TC0100000254.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100001713.mm.1 NR_040290.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100001713.mm.1 NR_040290.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100001713.mm.1 TC0100000254.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100001707.mm.1 NR_040290.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100001707.mm.1 NR_040290.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100001707.mm.1 TC0100000254.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100001708.mm.1 NR_040290.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100001708.mm.1 NR_040290.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100001708.mm.1 TC0100000254.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100001712.mm.1 NR_040290.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100001712.mm.1 NR_040290.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100001712.mm.1 TC0100000254.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100001714.mm.1 NR_040290.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100001714.mm.1 NR_040290.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100001714.mm.1 TC0100000254.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0