This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01229458_m1
See other IGF1OS GE Assays ›
Gene Symbol:
Igf1os
Gene Name:
insulin-like growth factor 1, opposite strand
Gene Aliases:
ENSMUSG00000058934, Gm10055, Gm16176, OTTMUSG00000031414
Chromosome Location:
Chr.10: 87826756 - 87864724 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01229458_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Igf1os
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA BC028536.1 - 1 - 2 95 1513858 79

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the BC028536.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Mus musculus Probe spans exons

MicroArray Probes:

Agilent Affymetrix

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2050365 BC028536.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2050366 AK051435.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2050366 BC028536.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 490432 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 815806 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 709062 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 371594 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 364002 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 289488 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 236177 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 124227 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 102432 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 960972 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 108003 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 448476 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 512195 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 536905 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 229493 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 399954 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 671447 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 813304 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 55590 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 281888 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 118206 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 168548 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 177468 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 36988 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P168479 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P119187 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2050365 BC028536.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2050366 AK051435.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2050366 BC028536.1 mouse_transcriptome
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07139724 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P168479 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P119187 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02180463 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07139703 - -
Whole Mouse Genome G4122A A_52_P411205 AK051435.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P411205 BC028536.1 mouse_transcriptome
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P119187 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P168479 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P07139703 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08746576 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27321915 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27322111 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27321964 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27321995 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08249074 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27321991 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27321911 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27321969 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27322024 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27322107 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08847587 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08760103 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27322113 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27322022 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27322033 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08916221 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08286749 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27322015 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08249072 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27321966 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27322109 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27321952 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27322038 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27321938 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27322036 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27321923 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27321949 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27322019 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27322110 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27322010 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27321954 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27321918 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27321946 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27321942 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08916216 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27322099 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27321980 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08786380 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27322039 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27321951 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27321986 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27322104 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08768759 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08760100 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27321992 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27321972 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27322009 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08768767 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27322029 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27322105 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08210688 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27321921 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08667575 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27321982 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27322034 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27321973 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27321994 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27321965 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27321971 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27321945 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27321998 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27322012 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08557995 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27321908 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27322004 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27321916 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27321941 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08672623 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08372807 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27321976 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27322017 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27322007 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08916222 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27321936 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08716393 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27321984 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27321979 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27321967 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27321961 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27322027 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27321988 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08760105 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27322008 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08880704 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08967976 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27322108 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27321925 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10371585 - -
Affymetrix Mouse Gene 1.0 ST Array 10371588 - -
Affymetrix Mouse Gene 1.0 ST Array 10371587 - -
Affymetrix HT MG-430 PM Array Plate 1428042_PM_at - -
Mouse Genome 430 2.0 Array 1428042_at - -
Mouse Genome 430 2.0 Array 1428042_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 627270_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 627270_PM_at - -
Affymetrix Mouse Clariom S Array PSR1000019932.mm.1 TC1000002635.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1000019930.mm.1 TC1000002635.mm.1 MTA TC
Mouse Expression Set 430 1428042_PM_at - -
Mouse Expression Set 430 1428042_at - -
Mouse Genome 430A 2.0 Array 1428042_PM_at - -
Mouse Genome 430A 2.0 Array 1428042_at - -
Affymetrix HT Mouse Genome 430A Array 1428042_PM_at - -
Affymetrix HT Mouse Genome 430A Array 1428042_at - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0