This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01229217_g1
See other IL2RG GE Assays ›
Gene Symbol:
Il2rg
Gene Name:
interleukin 2 receptor, gamma chain
Gene Aliases:
CD132, [g]c, gamma(c), gc, p64
Chromosome Location:
Chr.X: 101264387 - 101268206 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01229217_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Il2rg
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK143669.1 - 5 - 6 778 95
AK153873.1 - 5 - 6 793 95
BC014720.1 - 5 - 6 772 4924951 95
D13565.1 - 5 - 6 770 95
D13821.1 - 5 - 6 765 95
L20048.1 - 5 - 6 782 95
X75337.1 - 5 - 6 795 95

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0X00003437.mm.1

Set Membership:

Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

transmembrane signal receptor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0