This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01227236_gH
See other EIF5B GE Assays ›
Gene Symbol:
Eif5b
Gene Name:
eukaryotic translation initiation factor 5B
Gene Aliases:
A030003E17Rik, BC018347, IF2
Chromosome Location:
Chr.1: 37998000 - 38055579 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01227236_gH
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Eif5b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_198303.2 NP_938045.2 19 - 20 3279 76
GenBank mRNA AK037164.1 - 3 - 4 386 76
AK173016.1 - 3 - 4 352 76
BC055393.1 - 3 - 4 357 76
BC103806.1 - 18 - 19 4032 76

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0100000271.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Eif5b

Panther Classification:

Molecular Function -

translation initiation factor

Gene Ontology Categories:

Function(s) Process(es)

translation
translational initiation
regulation of translational initiation
nucleotide binding
translation initiation factor activity
GTPase activity
GTP binding
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100001955.mm.1 NM_198303.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100001955.mm.1 NM_198303.2 Refseq
Affymetrix Mouse Clariom S Array PSR0100001955.mm.1 TC0100000271.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0