This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01226610_g1
See other ACTR2 GE Assays ›
Gene Symbol:
Actr2
Gene Name:
ARP2 actin-related protein 2
Gene Aliases:
4921510D23Rik, AA409782, Arp2, D6Ertd746e
Chromosome Location:
Chr.11: 20062304 - 20112951 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01226610_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Actr2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_146243.2 NP_666355.1 1 - 2 148 71
GenBank mRNA AK032832.1 - 1 - 2 66 71
AK033014.1 - 1 - 2 66 71
AK088216.1 - 1 - 2 60 71
AK163217.1 - 1 - 2 61 71
BC027799.1 - 1 - 2 132 5325176 71
BC047530.1 - 1 - 2 47 71
BY278998.1 - 1 - 2 147 71

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Actr2

Panther Classification:

Molecular Function -

actin and actin related protein actin or actin-binding cytoskeletal protein cytoskeletal protein

Gene Ontology Categories:

Function(s) Process(es)

actin filament organization
establishment or maintenance of cell polarity
asymmetric cell division
meiotic chromosome movement towards spindle pole
cytosolic transport
actin cytoskeleton organization
meiotic cytokinesis
Arp2/3 complex-mediated actin nucleation
cilium assembly
meiotic cell cycle
spindle localization
nucleotide binding
actin binding
structural constituent of cytoskeleton
ATP binding
actin filament binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0