This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01226331_g1
See other DNAJC3 GE Assays ›
Gene Symbol:
Dnajc3
Gene Name:
DnaJ (Hsp40) homolog, subfamily C, member 3
Gene Aliases:
AA408985, AU067833, Dnajc3a, Dnajc3b, Prkri, mp58, p58, p58IPK
Chromosome Location:
Chr.14: 118937932 - 118981702 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01226331_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dnajc3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_008929.3 NP_032955.2 8 - 9 1116 83
GenBank mRNA AK004033.1 - 3 - 4 417 83
AK146097.1 - 8 - 9 1080 83
AK148270.1 - 8 - 9 1073 83
AK188318.1 - 2 - 3 191 83
BC013766.1 - 8 - 9 1061 2646147 83
U28423.1 - 8 - 9 1059 83

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1400001290.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Dnajc3

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of protein kinase activity
response to unfolded protein
endoplasmic reticulum unfolded protein response
negative regulation of apoptotic process
proteolysis involved in cellular protein catabolic process
negative regulation of endoplasmic reticulum stress-induced eIF2 alpha phosphorylation
protein kinase inhibitor activity
protein binding
chaperone binding
misfolded protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1400009722.mm.1 NM_008929.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400009722.mm.1 NM_008929.3 Refseq
Affymetrix Mouse Clariom S Array PSR1400009722.mm.1 TC1400001290.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0