This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01226326_g1
See other CLDN10 GE Assays ›
Gene Symbol:
Cldn10
Gene Name:
claudin 10
Gene Aliases:
6720456I16Rik, Cldn10a, Cldn10b, D14Ertd728e
Chromosome Location:
Chr.14: 118787871 - 118874525 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01226326_g1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cldn10
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001160096.1 NP_001153568.1 2 - 3 602 57
NM_001160097.1 NP_001153569.1 2 - 3 659 57
NM_001160098.1 NP_001153570.1 2 - 3 602 57
NM_001160099.1 NP_001153571.1 2 - 3 419 57
NM_021386.4 NP_067361.2 2 - 3 419 57
NM_023878.3 NP_076367.2 2 - 3 659 57
GenBank mRNA AF124425.1 - 2 - 3 388 57
AK020131.1 - 2 - 3 623 57
BC021770.1 - 2 - 3 612 5040100 57
BC029019.1 - 412 5356256 57
CA481582.1 - 2 - 3 610 57
CK332009.1 - 2 - 3 224 57
CK794879.1 - 2 - 3 419 57

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1400001288.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cldn10

Panther Classification:

Molecular Function -

tight junction

Gene Ontology Categories:

Function(s) Process(es)

transport
ion transport
structural molecule activity

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1400009706.mm.1 NM_001160096.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400009706.mm.1 NM_001160096.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400009706.mm.1 NM_001160097.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400009706.mm.1 NM_001160097.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400009706.mm.1 NM_001160098.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400009706.mm.1 NM_001160098.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400009706.mm.1 NM_001160099.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400009706.mm.1 NM_001160099.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400009706.mm.1 NM_021386.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400009706.mm.1 NM_021386.4 Refseq
Affymetrix Mouse Clariom S Array PSR1400009706.mm.1 NM_023878.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400009706.mm.1 NM_023878.3 Refseq
Affymetrix Mouse Clariom S Array PSR1400009706.mm.1 TC1400001288.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1400009708.mm.1 NM_001160096.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400009708.mm.1 NM_001160096.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400009708.mm.1 NM_001160097.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400009708.mm.1 NM_001160097.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400009708.mm.1 NM_001160098.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400009708.mm.1 NM_001160098.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400009708.mm.1 NM_001160099.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400009708.mm.1 NM_001160099.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400009708.mm.1 NM_021386.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400009708.mm.1 NM_021386.4 Refseq
Affymetrix Mouse Clariom S Array PSR1400009708.mm.1 NM_023878.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400009708.mm.1 NM_023878.3 Refseq
Affymetrix Mouse Clariom S Array PSR1400009708.mm.1 TC1400001288.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 348339 - -
ABI Mouse Genome Survey Microarray 362550 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0