This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01224431_g1
See other EDNRB GE Assays ›
Gene Symbol:
Ednrb
Gene Name:
endothelin receptor type B
Gene Aliases:
ET-B, ET-BR, ETR-b, ETb, Sox10m1
Chromosome Location:
Chr.14: 103814615 - 103844476 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01224431_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ednrb
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001136061.2 NP_001129533.1 1 - 2 350 59
GenBank mRNA AK082103.1 - 1 - 2 151 59
AK083415.1 - 1 - 2 47 59
BC026553.1 - 1 - 2 276 4971909 59

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK082103.1, AK083415.1, BC026553.1, NM_001136061.2 transcripts.

Set Membership:

Amplicon length less than or equal to 70 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
neural crest cell migration
positive regulation of protein phosphorylation
regulation of pH
signal transduction
G-protein coupled receptor signaling pathway
phospholipase C-activating G-protein coupled receptor signaling pathway
positive regulation of cytosolic calcium ion concentration
peripheral nervous system development
posterior midgut development
regulation of blood pressure
positive regulation of cell proliferation
negative regulation of neuron maturation
vein smooth muscle contraction
sensory perception of pain
cGMP-mediated signaling
melanocyte differentiation
regulation of fever generation
negative regulation of cellular protein metabolic process
response to lipopolysaccharide
enteric smooth muscle cell differentiation
positive regulation of urine volume
positive regulation of renal sodium excretion
epithelial fluid transport
vasoconstriction
vasodilation
negative regulation of apoptotic process
pigmentation
developmental pigmentation
macrophage chemotaxis
response to pain
enteric nervous system development
regulation of epithelial cell proliferation
regulation of sensory perception of pain
positive regulation of penile erection
endothelin receptor signaling pathway
signal transducer activity
G-protein coupled receptor activity
endothelin receptor activity
protein binding
peptide hormone binding
type 1 angiotensin receptor binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1400019790.mm.1 NM_001136061.2 Refseq
Affymetrix Mouse Clariom S Array PSR1400019790.mm.1 TC1400002641.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1400019785.mm.1 NM_001136061.2 Refseq
Affymetrix Mouse Clariom S Array PSR1400019785.mm.1 NM_001276296.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400019785.mm.1 NM_007904.4 Refseq
Affymetrix Mouse Clariom S Array PSR1400019785.mm.1 TC1400002641.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1400019788.mm.1 NM_001136061.2 Refseq
Affymetrix Mouse Clariom S Array PSR1400019788.mm.1 TC1400002641.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1400019789.mm.1 NM_001136061.2 Refseq
Affymetrix Mouse Clariom S Array PSR1400019789.mm.1 TC1400002641.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 499502 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-655dcf6cc8-78hlp:80/100.66.128.142:80.
git-commit: 0d764ad91c70359c551b4d51a80efc6847a0ee54
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0