This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01223055_m1
See other PTGDR2 GE Assays ›
Gene Symbol:
Ptgdr2
Gene Name:
prostaglandin D2 receptor 2
Gene Aliases:
Crth2, Gpr44, Grp45
Chromosome Location:
Chr.19: 10927861 - 10942511 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01223055_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ptgdr2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_009962.3 NP_034092.1 2 - 3 193 63
GenBank mRNA AB109092.1 - 2 - 3 193 63
AK041947.1 - 2 - 3 191 63
BC111906.1 - 2 - 3 173 40039170 63

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AB109092.1, BC111906.1, NM_009962.3 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ptgdr2

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

chemotaxis
signal transduction
G-protein coupled receptor signaling pathway
adenylate cyclase-inhibiting G-protein coupled receptor signaling pathway
neuropeptide signaling pathway
chemical synaptic transmission
calcium-mediated signaling
positive regulation of G-protein coupled receptor protein signaling pathway
negative regulation of male germ cell proliferation
prostaglandin J receptor activity
signal transducer activity
G-protein coupled receptor activity
prostaglandin D receptor activity
prostaglandin F receptor activity
neuropeptide binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1900002683.mm.1 NM_009962.3 Refseq
Affymetrix Mouse Clariom S Array PSR1900002683.mm.1 TC1900000213.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0