This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01222980_m1
See other RMDN1 GE Assays ›
Gene Symbol:
Rmdn1
Gene Name:
regulator of microtubule dynamics 1
Gene Aliases:
Fam82b, RMD-1
Chromosome Location:
Chr.4: 19574862 - 19606932 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01222980_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rmdn1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_025476.6 NP_079752.3 1 - 2 260 142
GenBank mRNA AK002442.1 - 1 - 2 143 142
AK009059.1 - 1 - 2 117 142
AK010421.2 - 1 - 2 137 142
AK029446.1 - 1 - 2 120 142
BC025158.1 - 1 - 2 133 4023319 142
BC096046.1 - 1 - 2 237 1397855 142

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Rmdn1

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0