This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01222274_m1
See other POLB GE Assays ›
Gene Symbol:
Polb
Gene Name:
polymerase (DNA directed), beta
Gene Aliases:
A430088C08Rik
Chromosome Location:
Chr.8: 22628119 - 22653468 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01222274_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Polb
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_011130.2 NP_035260.1 6 - 7 438 89
GenBank mRNA AF513911.1 - 6 - 7 415 89
AK077127.1 - 6 - 7 423 89
AK146745.1 - 6 - 7 427 89
AK151436.1 - 6 - 7 423 89
AV488131.1 - 6 - 7 436 89
BC060998.1 - 6 - 7 419 6774075 89

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0800001872.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Polb

Panther Classification:

Molecular Function -

DNA-directed DNA polymerase

Gene Ontology Categories:

Function(s) Process(es)

DNA replication
DNA repair
base-excision repair
base-excision repair, gap-filling
pyrimidine dimer repair
apoptotic process
inflammatory response
cellular response to DNA damage stimulus
salivary gland morphogenesis
intrinsic apoptotic signaling pathway in response to DNA damage
somatic diversification of immunoglobulins
somatic hypermutation of immunoglobulin genes
lymph node development
spleen development
homeostasis of number of cells
neuron apoptotic process
immunoglobulin heavy chain V-D-J recombination
DNA biosynthetic process
DNA binding
damaged DNA binding
DNA-directed DNA polymerase activity
microtubule binding
transferase activity
nucleotidyltransferase activity
lyase activity
enzyme binding
DNA polymerase activity
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0800014786.mm.1 NM_011130.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800014786.mm.1 NM_011130.2 Refseq
Affymetrix Mouse Clariom S Array PSR0800014786.mm.1 TC0800001872.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header