This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01221930_m1
See other OGDHL GE Assays ›
Gene Symbol:
Ogdhl
Gene Name:
oxoglutarate dehydrogenase-like
Gene Aliases:
-
Chromosome Location:
Chr.14: 32322012 - 32348150 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01221930_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ogdhl
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001081130.1 NP_001074599.1 8 - 9 1123 103
GenBank mRNA BC157970.1 - 7 - 8 923 9007292 103
BC157971.1 - 7 - 8 923 9007293 103

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1400000344.mm.1

Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

dehydrogenase

Gene Ontology Categories:

Function(s) Process(es)

tricarboxylic acid cycle
biological_process
molecular_function
oxoglutarate dehydrogenase (succinyl-transferring) activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1400003038.mm.1 NM_001081130.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400003038.mm.1 NM_001081130.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400003038.mm.1 TC1400000344.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0