This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01221713_g1
See other PRR14L GE Assays ›
Gene Symbol:
Prr14l
Gene Name:
proline rich 14-like
Gene Aliases:
6030436E02Rik, C330019G07Rik, Prl14l
Chromosome Location:
Chr.5: 32789206 - 32854338 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01221713_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Prr14l
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_194340.2 NP_919321.2 8 - 9 5743 107
GenBank mRNA AK049285.1 - 1 - 2 75 107
AY372183.1 - 3 - 4 964 107
BC059908.1 - 1 - 2 175 107
BC094374.1 - 3 - 4 1290 30531815 107

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Prr14l

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500020138.mm.1 NM_194340.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500020138.mm.1 NM_194340.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500020138.mm.1 TC0500003726.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0