This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01220978_g1
See other TUBGCP3 GE Assays ›
Gene Symbol:
Tubgcp3
Gene Name:
tubulin, gamma complex associated protein 3
Gene Aliases:
GCP3, Spc98p
Chromosome Location:
Chr.8: 12614277 - 12672243 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01220978_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tubgcp3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_198031.1 NP_932148.1 9 - 10 1101 88
GenBank mRNA AK051775.1 - 9 - 10 1113 88
AK150149.1 - 9 - 10 1125 88
AK165503.1 - 9 - 10 1124 88
AK171491.1 - 9 - 10 1107 88
AK171804.1 - 9 - 10 1116 88
BC025647.1 - 4 - 5 348 88
BC058566.1 - 9 - 10 1101 6466057 88

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tubgcp3

Panther Classification:

Molecular Function -

non-motor microtubule binding protein

Gene Ontology Categories:

Function(s) Process(es)

microtubule cytoskeleton organization
microtubule nucleation
biological_process
gamma-tubulin binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0