This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01220863_m1
See other SDCBP GE Assays ›
Gene Symbol:
Sdcbp
Gene Name:
syndecan binding protein
Gene Aliases:
MDA-9, Sycl, syntenin-1
Chromosome Location:
Chr.4: 6365680 - 6396122 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01220863_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Sdcbp
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001098227.1 NP_001091697.1 2 - 3 147 132
NM_016807.2 NP_058087.2 2 - 3 147 132
GenBank mRNA AF077527.1 - 2 - 3 106 132
AK014678.1 - 121 132
AK028429.1 - 2 - 3 120 132
AK033412.1 - 2 - 3 145 132
AK146085.1 - 2 - 3 119 132
AK150278.1 - 2 - 3 112 132
AK150321.1 - 2 - 3 115 132
AK150544.1 - 2 - 3 119 132
AK150623.1 - 2 - 3 123 132
AK150661.1 - 2 - 3 115 132
AK150707.1 - 2 - 3 118 132
AK150721.1 - 2 - 3 127 132
AK150736.1 - 2 - 3 115 132
AK150801.1 - 2 - 3 117 132
AK150996.1 - 2 - 3 119 132
AK151050.1 - 2 - 3 119 132
AK152001.1 - 2 - 3 118 132
AK152017.1 - 2 - 3 119 132
AK152080.1 - 2 - 3 126 132
AK152109.1 - 2 - 3 119 132
AK152434.1 - 2 - 3 119 132
AK152552.1 - 2 - 3 123 132
AK152577.1 - 2 - 3 118 132
AK152855.1 - 2 - 3 119 132
AK153042.1 - 2 - 3 119 132
AK153112.1 - 2 - 3 123 132
AK159124.1 - 2 - 3 115 132
AK159538.1 - 2 - 3 120 132
AK159677.1 - 2 - 3 126 132
AK165657.1 - 2 - 3 117 132
AK167513.1 - 2 - 3 115 132
AK168004.1 - 2 - 3 122 132
AK168777.1 - 2 - 3 115 132
AK169219.1 - 2 - 3 124 132
AK169415.1 - 2 - 3 153 132
AK169687.1 - 2 - 3 119 132
AK171162.1 - 2 - 3 119 132
BC019400.1 - 2 - 3 110 4017442 131
BY779386.1 - 2 - 3 148 132

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AF077527.1, AK028429.1, AK033412.1, AK146085.1, AK150278.1, AK150321.1, AK150544.1, AK150623.1, AK150661.1, AK150707.1, AK150721.1, AK150736.1, AK150801.1, AK150996.1, AK151050.1, AK152001.1, AK152017.1, AK152109.1, AK152434.1, AK152552.1, AK152577.1, AK152855.1, AK153042.1, AK153112.1, AK159538.1, AK159677.1, AK165657.1, AK167513.1, AK168004.1, AK168777.1, AK169219.1, AK169415.1, AK169687.1, AK171162.1, BC019400.1, NM_001098227.1, NM_016807.2 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Sdcbp

Panther Classification:

Molecular Function -

membrane trafficking regulatory protein

Gene Ontology Categories:

Function(s) Process(es)

Ras protein signal transduction
positive regulation of phosphorylation
positive regulation of exosomal secretion
positive regulation of extracellular exosome assembly
glycoprotein binding
frizzled binding
interleukin-5 receptor binding
protein binding
protein C-terminus binding
growth factor binding
neurexin family protein binding
identical protein binding
protein homodimerization activity
syndecan binding
ephrin receptor binding
protein heterodimerization activity
protein N-terminus binding
cell adhesion molecule binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0400000264.mm.1 NM_001098227.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400000264.mm.1 NM_001098227.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400000264.mm.1 NM_016807.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400000264.mm.1 NM_016807.2 Refseq
Affymetrix Mouse Clariom S Array PSR0400000264.mm.1 TC0400000039.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0