Assay Details
Gene Symbol: Unc5d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153135.3 | NP_694775.1 | 3 - 4 | 754 | 77 | |
GenBank mRNA | AJ487854.1 | - | 3 - 4 | 755 | 77 | |
BC137615.1 | - | 3 - 4 | 545 | 8860634 | 77 | |
BC144783.1 | - | 3 - 4 | 545 | 9053331 | 77 | |
BC144784.1 | - | 3 - 4 | 545 | 9053332 | 77 | |
BC145695.1 | - | 3 - 4 | 545 | 40130998 | 77 | |
FJ824810.1 | - | 3 - 4 | 687 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
