This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01220266_m1
See other LCORL GE Assays ›
Gene Symbol:
Lcorl
Gene Name:
ligand dependent nuclear receptor corepressor-like
Gene Aliases:
Mlr1
Chromosome Location:
Chr.5: 45697184 - 45860115 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01220266_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Lcorl
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001163073.1 NP_001156545.1 4 - 5 476 85
NM_172153.3 NP_742165.1 4 - 5 489 85
NM_178142.5 NP_835278.2 4 - 5 476 85
GenBank mRNA AB076078.1 - 4 - 5 488 85
AK041987.1 - 4 - 5 435 85
AK042567.1 - 4 - 5 490 85
AK043498.1 - 4 - 5 434 85
AK043845.1 - 4 - 5 444 85
AK154398.1 - 4 - 5 437 85
AK155421.1 - 4 - 5 437 85
BC066151.1 - 4 - 5 406 85
BC138563.1 - 4 - 5 476 8861583 85

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500002346.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Lcorl

Panther Classification:

Molecular Function -

DNA-binding transcription factor gene-specific transcriptional regulator

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
DNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500021528.mm.1 NM_001163073.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500021528.mm.1 NM_001163073.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500021528.mm.1 NM_178142.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500021528.mm.1 NM_178142.5 Refseq
Affymetrix Mouse Clariom S Array PSR0500021528.mm.1 TC0500002346.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500021523.mm.1 NM_001163073.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500021523.mm.1 NM_001163073.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500021523.mm.1 NM_172153.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500021523.mm.1 NM_172153.3 Refseq
Affymetrix Mouse Clariom S Array PSR0500021523.mm.1 NM_178142.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500021523.mm.1 NM_178142.5 Refseq
Affymetrix Mouse Clariom S Array PSR0500021523.mm.1 TC0500002346.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500021530.mm.1 NM_001163073.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500021530.mm.1 NM_001163073.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500021530.mm.1 NM_178142.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500021530.mm.1 NM_178142.5 Refseq
Affymetrix Mouse Clariom S Array PSR0500021530.mm.1 TC0500002346.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500021520.mm.1 NM_001163073.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500021520.mm.1 NM_001163073.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500021520.mm.1 NM_172153.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500021520.mm.1 NM_172153.3 Refseq
Affymetrix Mouse Clariom S Array PSR0500021520.mm.1 NM_178142.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500021520.mm.1 NM_178142.5 Refseq
Affymetrix Mouse Clariom S Array PSR0500021520.mm.1 TC0500002346.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56ffd697dc-88d4p:80/100.66.130.94:80.
git-commit: 082a6d415be982b1f92c35e55dd71609d4576d3e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0