Assay Details
| Gene Symbol: S100a13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009113.4 | NP_033139.2 | 2 - 3 | 298 | 61 | |
| GenBank mRNA | AK002755.1 | - | 1 - 2 | 97 | 61 | |
| AK008895.1 | - | 1 - 2 | 97 | 61 | ||
| BC005687.1 | - | 2 - 3 | 298 | 3155915 | 61 | |
| CK334302.1 | - | 1 - 2 | 265 | 61 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK002755.1, AK008895.1, BC005687.1 transcripts.
Set Membership: |
|

S100a13
Genomic Map