This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01220067_g1
See other S100A13 GE Assays ›
Gene Symbol:
S100a13
Gene Name:
S100 calcium binding protein A13
Gene Aliases:
-
Chromosome Location:
Chr.3: 90514435 - 90524581 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01220067_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: S100a13
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_009113.4 NP_033139.2 1 - 2 37 62
GenBank mRNA BC005687.1 - 1 - 2 37 3155915 62

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the BC005687.1, NM_009113.4 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene S100a13

Panther Classification:

Molecular Function -

calmodulin-related

Gene Ontology Categories:

Function(s) Process(es)

transport
positive regulation of cell proliferation
regulation of cell shape
protein transport
positive regulation of I-kappaB kinase/NF-kappaB signaling
cytokine secretion
interleukin-1 alpha secretion
copper ion binding
calcium ion binding
zinc ion binding
lipid binding
fibroblast growth factor binding
protein homodimerization activity
metal ion binding
RAGE receptor binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0300006243.mm.1 NM_009113.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300006243.mm.1 NM_009113.4 Refseq
Affymetrix Mouse Clariom S Array PSR0300006243.mm.1 TC0300000800.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300006244.mm.1 NM_009113.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300006244.mm.1 NM_009113.4 Refseq
Affymetrix Mouse Clariom S Array PSR0300006244.mm.1 TC0300000800.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 908701 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0