This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01220053_g1
See other DENND4B GE Assays ›
Gene Symbol:
Dennd4b
Gene Name:
DENN/MADD domain containing 4B
Gene Aliases:
-
Chromosome Location:
Chr.3: 90264920 - 90280665 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01220053_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dennd4b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_201407.4 NP_958809.3 6 - 7 1229 84
GenBank mRNA AK155638.1 - 6 - 7 1211 84
AY512564.1 - 4 - 5 837 84
BC066102.1 - 5 - 6 1008 30531856 84
BC151194.1 - 5 - 6 875 9088082 84

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0300000787.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Dennd4b

Gene Ontology Categories:

Function(s) Process(es)

regulation of Rab protein signal transduction
positive regulation of GTPase activity
guanyl-nucleotide exchange factor activity
Rab guanyl-nucleotide exchange factor activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0300006150.mm.1 NM_201407.4 Refseq
Affymetrix Mouse Clariom S Array PSR0300006150.mm.1 TC0300000787.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300006151.mm.1 NM_201407.4 Refseq
Affymetrix Mouse Clariom S Array PSR0300006151.mm.1 TC0300000787.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0