This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01219898_m1
See other MORC3 GE Assays ›
Gene Symbol:
Morc3
Gene Name:
microrchidia 3
Gene Aliases:
1110051N18Rik, AI452146, BF318192, D16Jhu32e, NXP2, Zcwcc3
Chromosome Location:
Chr.16: 93832121 - 93876077 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01219898_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Morc3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001045529.3 NP_001038994.2 13 - 14 1654 77
GenBank mRNA AK132226.1 - 12 - 13 1614 77
AK168211.1 - 4 - 5 341 77
AK172896.1 - 13 - 14 1654 77
BC098072.1 - 13 - 14 1495 30109159 77
BC145705.1 - 13 - 14 1603 40131008 77

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1600001048.mm.1

Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

protein phosphorylation
cell aging
post-embryonic development
peptidyl-serine phosphorylation
negative regulation of fibroblast proliferation
protein stabilization
maintenance of protein location in nucleus
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1600008164.mm.1 NM_001045529.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600008164.mm.1 NM_001045529.3 Refseq
Affymetrix Mouse Clariom S Array PSR1600008164.mm.1 TC1600001048.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1600008163.mm.1 NM_001045529.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600008163.mm.1 NM_001045529.3 Refseq
Affymetrix Mouse Clariom S Array PSR1600008163.mm.1 TC1600001048.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0