This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01219210_m1
See other 4930557J02RIK GE Assays ›
Gene Symbol:
4930557J02Rik
Gene Name:
RIKEN cDNA 4930557J02 gene
Gene Aliases:
-
Chromosome Location:
Chr.5: 34118740 - 34135339 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01219210_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 4930557J02Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_040703.1 3 - 4 925 63
GenBank mRNA AK016164.1 - 3 - 4 927 63

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream DNA polymerase N Poln POL4P Anti-sense Chr. 5 - 34007179 - 34169526 on NCBI Build GRCm38
Downstream HAUS augmin-like complex,
subunit 3
Haus3 D4S43h,
D5H4S43,
D5H4S43E
18.6kb Anti-sense Chr. 5 - 34153896 - 34169424 on NCBI Build GRCm38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500000368.mm.1

Set Membership:

Amplicon length less than or equal to 70 3' Most Mus musculus 1700 Probe spans exons

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2337133 AK016164.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P106394 - -
Array Name Probe IDs Targets Target Type
Affymetrix HT MG-430 PM Array Plate 1454189_PM_at - -
Mouse Genome 430 2.0 Array 1454189_at - -
Mouse Genome 430 2.0 Array 1454189_PM_at - -
Affymetrix Mouse Clariom S Array PSR0500003518.mm.1 NR_040703.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500003518.mm.1 NR_040703.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500003518.mm.1 TC0500000368.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500003519.mm.1 NR_040703.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500003519.mm.1 NR_040703.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500003519.mm.1 TC0500000368.mm.1 MTA TC
Mouse Expression Set 430 1454189_PM_at - -
Mouse Expression Set 430 1454189_at - -
Affymetrix HT Mouse Genome 430B Array 1454189_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1454189_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 818489 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 100050239 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0