This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01219113_g1
See other 1700029J03RIK GE Assays ›
Gene Symbol:
1700029J03Rik
Gene Name:
RIKEN cDNA 1700029J03 gene
Gene Aliases:
-
Chromosome Location:
Chr.16: 93396815 - 93458872 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01219113_g1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 1700029J03Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_040494.1 2 - 3 481 80
GenBank mRNA AK006500.1 - 2 - 3 482 80
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BY706535.1 - 2 - 3 482 Chr. 16 - 93396918 - 93458872 [-] on Build GRCm38 80

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream microRNA 802 Mir802 Mirn802,
mir-802,
mmu-mir-802
-27.0kb Anti-sense Chr. 16 - 93369720 - 93369816 on NCBI Build GRCm38
Downstream predicted gene,
30695
Gm30695 88.9kb Sense Chr. 16 - 93486513 - 93531385 on NCBI Build GRCm38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1600002104.mm.1

Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus 1700 Probe spans exons

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 738611 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 152210 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 360093 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 692261 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 378375 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 290800 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 714482 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 275066 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 736341 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 129648 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 898811 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 646322 AK038647.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 632620 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 538658 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 803268 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 202525 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 896841 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 838597 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 812347 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 478420 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 559048 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 591860 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 234976 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 800639 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 108927 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 35517 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 142998 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 129742 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08688117 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08688051 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03259823 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08688014 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P08688051 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30975854 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30975845 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30975511 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12513445 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30975549 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30975562 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P13015793 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30975535 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30975851 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30975859 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30975885 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30975537 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12493743 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30975567 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30975441 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30975848 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30975577 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30975550 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30975846 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12977405 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30975452 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P13015798 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30975580 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30975891 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30975582 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30975517 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12948262 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30975540 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30975538 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12499375 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30975843 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30975894 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30975844 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30975571 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30975557 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12513444 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30975893 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P13041451 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12513447 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30975527 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12499376 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30975855 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30975440 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30975564 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12684133 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30975516 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30975544 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30975882 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30975853 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30975575 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30975896 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30975847 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30975448 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30975884 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30975542 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30975897 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30975530 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12540827 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30975841 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30975881 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12684119 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12482061 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30975536 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P13054326 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30975445 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30975509 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30975849 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30975518 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30975560 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30975450 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30975555 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12684131 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30975533 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30975552 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30975840 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12493741 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12875324 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12977418 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30975547 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30975451 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1600016165.mm.1 NR_040494.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600016165.mm.1 NR_040494.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600016165.mm.1 TC1600002104.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 555176 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 102030152 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0