Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0500033725.mm.1 |
NM_201369.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500033725.mm.1 |
NM_201369.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500033725.mm.1 |
TC0500003688.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500033724.mm.1 |
NM_201369.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500033724.mm.1 |
NM_201369.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500033724.mm.1 |
TC0500003688.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500033721.mm.1 |
NM_201369.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500033721.mm.1 |
NM_201369.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500033721.mm.1 |
TC0500003688.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500033722.mm.1 |
NM_201369.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500033722.mm.1 |
NM_201369.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500033722.mm.1 |
TC0500003688.mm.1 |
MTA TC |