This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01218344_m1
See other 1700025M24RIK GE Assays ›
Gene Symbol:
1700025M24Rik
Gene Name:
RIKEN cDNA 1700025M24 gene
Gene Aliases:
-
Chromosome Location:
Chr.5: 73268580 - 73284184 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01218344_m1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 1700025M24Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_040687.1 2 - 3 111 69
GenBank mRNA BC048655.1 - 2 - 3 108 69
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA CA465171.1 - 111 Chr. - - [-] on Build 69

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream furry homolog-like (Drosophila) Fryl 2010313D22Rik,
2310004H21Rik,
2510002A14Rik,
9030227G01Rik,
mKIAA0826
-25.4kb Sense Chr. 5 - 73020191 - 73256645 on NCBI Build GRCm38
Downstream OCIA domain containing 1 Ociad1 6030432N09Rik,
AI481327,
AW557942,
Asrij,
B230209J16Rik,
BB021357,
Emi2,
Imi2,
OCIA,
TPA018
9.7kb Anti-sense Chr. 5 - 73292772 - 73314077 on NCBI Build GRCm38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500002590.mm.1

Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500022988.mm.1 NR_040687.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500022988.mm.1 NR_040687.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500022988.mm.1 NR_040688.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500022988.mm.1 NR_040688.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500022988.mm.1 TC0500002590.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500022987.mm.1 NR_040687.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500022987.mm.1 NR_040687.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500022987.mm.1 TC0500002590.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500022990.mm.1 NR_040687.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500022990.mm.1 NR_040687.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500022990.mm.1 NR_040688.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500022990.mm.1 NR_040688.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500022990.mm.1 NR_040689.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500022990.mm.1 NR_040689.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500022990.mm.1 TC0500002590.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0