This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01218334_g1
See other SEMA4A GE Assays ›
Gene Symbol:
Sema4a
Gene Name:
sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and short cytoplasmic domain, (semaphorin) 4A
Gene Aliases:
AI132332, SemB, Semab
Chromosome Location:
Chr.3: 88435959 - 88461182 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01218334_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Sema4a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001163489.1 NP_001156961.1 8 - 9 876 60
NM_001163490.1 NP_001156962.1 8 - 9 955 60
NM_001163491.1 NP_001156963.1 8 - 9 914 60
NM_013658.3 NP_038686.3 8 - 9 1054 60
GenBank mRNA AK035918.1 - 7 - 8 916 60
AK077976.1 - 8 - 9 1034 60
AK152728.1 - 8 - 9 1046 60
AK153164.1 - 8 - 9 1046 60
AK154392.1 - 8 - 9 1015 60
BC025800.1 - 8 - 9 972 5102015 60
X85991.1 - 8 - 9 876 60

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0300002312.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Sema4a

Panther Classification:

Molecular Function -

membrane-bound signaling molecule intercellular signal molecule

Gene Ontology Categories:

Function(s)

angiogenesis
T cell differentiation involved in immune response
immune system process
multicellular organism development
nervous system development
axonogenesis
regulation of cell shape
regulation of endothelial cell migration
negative regulation of angiogenesis
cell differentiation
T-helper 1 cell differentiation
semaphorin-plexin signaling pathway

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0300017543.mm.1 NM_001163489.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300017543.mm.1 NM_001163489.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300017543.mm.1 NM_001163490.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300017543.mm.1 NM_001163490.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300017543.mm.1 NM_001163491.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300017543.mm.1 NM_001163491.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300017543.mm.1 NM_013658.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300017543.mm.1 NM_013658.3 Refseq
Affymetrix Mouse Clariom S Array PSR0300017543.mm.1 TC0300002312.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300017540.mm.1 NM_001163489.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300017540.mm.1 NM_001163489.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300017540.mm.1 NM_001163490.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300017540.mm.1 NM_001163490.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300017540.mm.1 NM_001163491.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300017540.mm.1 NM_001163491.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300017540.mm.1 NM_013658.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300017540.mm.1 NM_013658.3 Refseq
Affymetrix Mouse Clariom S Array PSR0300017540.mm.1 TC0300002312.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300017541.mm.1 NM_001163489.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300017541.mm.1 NM_001163489.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300017541.mm.1 NM_001163490.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300017541.mm.1 NM_001163490.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300017541.mm.1 NM_001163491.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300017541.mm.1 NM_001163491.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300017541.mm.1 NM_013658.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300017541.mm.1 NM_013658.3 Refseq
Affymetrix Mouse Clariom S Array PSR0300017541.mm.1 TC0300002312.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0