This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01218329_m1
See other SEMA4A GE Assays ›
Gene Symbol:
Sema4a
Gene Name:
sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and short cytoplasmic domain, (semaphorin) 4A
Gene Aliases:
AI132332, SemB, Semab
Chromosome Location:
Chr.3: 88435959 - 88461182 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01218329_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Sema4a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001163489.1 NP_001156961.1 11 - 12 1382 131
NM_001163490.1 NP_001156962.1 11 - 12 1461 131
NM_001163491.1 NP_001156963.1 11 - 12 1420 131
NM_013658.3 NP_038686.3 11 - 12 1560 131
GenBank mRNA AK035918.1 - 10 - 11 1422 131
AK077976.1 - 11 - 12 1540 132
AK152728.1 - 11 - 12 1552 131
AK153164.1 - 11 - 12 1552 131
AK154392.1 - 11 - 12 1521 131
BC025800.1 - 11 - 12 1478 5102015 131

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Sema4a

Panther Classification:

Molecular Function -

membrane-bound signaling molecule intercellular signal molecule

Gene Ontology Categories:

Function(s)

angiogenesis
T cell differentiation involved in immune response
immune system process
multicellular organism development
nervous system development
axonogenesis
regulation of cell shape
regulation of endothelial cell migration
negative regulation of angiogenesis
cell differentiation
T-helper 1 cell differentiation
semaphorin-plexin signaling pathway

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0