Assay Details
Gene Symbol: Pmf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF348509.1 | - | 1 - 2 | 171 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() |

Gene Symbol: Pmf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF348509.1 | - | 1 - 2 | 171 | 68 |
Set Membership: |
![]() ![]() ![]() |