This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01218200_m1
See other S100A7A GE Assays ›
Gene Symbol:
S100a7a
Gene Name:
S100 calcium binding protein A7A
Gene Aliases:
AY465109, Gm1020, S100A7L1, S100A7f, S100a15, S100a15a, S100a17l1
Chromosome Location:
Chr.3: 90651934 - 90658130 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01218200_m1
Size
Availability Inventoried
Catalog # 4331182
Price 244.00
Your Price
Online offer: 200.00
200.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: S100a7a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_199422.1 NP_955454.1 1 - 2 44 105
GenBank mRNA AY465109.1 - 1 - 2 44 105

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AY465109.1, NM_199422.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene S100a7a

Panther Classification:

Molecular Function -

calmodulin-related

Gene Ontology Categories:

Function(s) Process(es)

inflammatory response
cytokine metabolic process
positive chemotaxis
calcium ion binding
protein binding
chemoattractant activity
protein self-association
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0300006315.mm.1 NM_199422.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300006315.mm.1 NM_199422.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300006315.mm.1 TC0300000810.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0