This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01217908_g1
See other NOC3L GE Assays ›
Gene Symbol:
Noc3l
Gene Name:
nucleolar complex associated 3 homolog (S. cerevisiae)
Gene Aliases:
AF233884, Fad24
Chromosome Location:
Chr.19: 38788128 - 38819270 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01217908_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Noc3l
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_021315.2 NP_067290.2 14 - 15 1778 121
GenBank mRNA AB077991.1 - 14 - 15 1741 121
AF233884.1 - 3 - 4 241 121
AK134051.1 - 14 - 15 1737 121
AK168944.1 - 14 - 15 1781 121
BC031132.1 - 14 - 15 1741 4954951 121

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1900001472.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Noc3l

Gene Ontology Categories:

Function(s) Process(es)

fat cell differentiation
chromatin binding
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1900012829.mm.1 NM_021315.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900012829.mm.1 NM_021315.2 Refseq
Affymetrix Mouse Clariom S Array PSR1900012829.mm.1 TC1900001472.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1900012828.mm.1 NM_021315.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900012828.mm.1 NM_021315.2 Refseq
Affymetrix Mouse Clariom S Array PSR1900012828.mm.1 TC1900001472.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0