Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0500020356.mm.1 |
NM_001289803.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500020356.mm.1 |
NM_001289804.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500020356.mm.1 |
NM_181857.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500020356.mm.1 |
TC0500002229.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500020358.mm.1 |
NM_001289803.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500020358.mm.1 |
NM_001289804.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500020358.mm.1 |
NM_181857.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500020358.mm.1 |
TC0500002229.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500020351.mm.1 |
NM_001289803.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500020351.mm.1 |
NM_001289804.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500020351.mm.1 |
NM_181857.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500020351.mm.1 |
TC0500002229.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500020353.mm.1 |
NM_001289803.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500020353.mm.1 |
NM_001289804.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500020353.mm.1 |
NM_181857.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500020353.mm.1 |
TC0500002229.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500020354.mm.1 |
NM_001289803.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500020354.mm.1 |
NM_001289804.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500020354.mm.1 |
NM_181857.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500020354.mm.1 |
TC0500002229.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500020357.mm.1 |
NM_001289803.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500020357.mm.1 |
NM_001289804.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500020357.mm.1 |
NM_181857.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500020357.mm.1 |
TC0500002229.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500020352.mm.1 |
NM_001289803.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500020352.mm.1 |
NM_001289804.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500020352.mm.1 |
NM_181857.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500020352.mm.1 |
TC0500002229.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500020355.mm.1 |
NM_001289803.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500020355.mm.1 |
NM_001289804.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500020355.mm.1 |
NM_181857.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500020355.mm.1 |
TC0500002229.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500020375.mm.1 |
NM_001289803.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500020375.mm.1 |
NM_001289804.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500020375.mm.1 |
NM_181857.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500020375.mm.1 |
TC0500002229.mm.1 |
MTA TC |