Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0800001213.mm.1 |
NM_027276.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800001213.mm.1 |
NM_027276.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800001213.mm.1 |
TC0800000130.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800001217.mm.1 |
NM_027276.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800001217.mm.1 |
NM_027276.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800001217.mm.1 |
TC0800000130.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800001219.mm.1 |
NM_027276.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800001219.mm.1 |
NM_027276.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800001219.mm.1 |
TC0800000130.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800001214.mm.1 |
NM_027276.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800001214.mm.1 |
NM_027276.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800001214.mm.1 |
TC0800000130.mm.1 |
MTA TC |