This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01217129_m1
See other CDC16 GE Assays ›
Gene Symbol:
Cdc16
Gene Name:
CDC16 cell division cycle 16
Gene Aliases:
2700071J12Rik, 2810431D22Rik, APC6
Chromosome Location:
Chr.8: 13757646 - 13781938 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01217129_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cdc16
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_027276.2 NP_081552.2 3 - 4 313 85
GenBank mRNA AK147938.1 - 3 - 4 328 85
AK158014.1 - 4 - 5 586 85
AK167965.1 - 3 - 4 328 85
AK168978.1 - 3 - 4 300 85
BC026606.1 - 3 - 4 313 4976424 85

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK158014.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cdc16

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Gene Ontology Categories:

Function(s) Process(es)

cell cycle
mitotic nuclear division
cell division
protein K11-linked ubiquitination
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0800001221.mm.1 NM_027276.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800001221.mm.1 NM_027276.2 Refseq
Affymetrix Mouse Clariom S Array PSR0800001221.mm.1 TC0800000130.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0