This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01216812_m1
See other S100A16 GE Assays ›
Gene Symbol:
S100a16
Gene Name:
S100 calcium binding protein A16
Gene Aliases:
2300002L21Rik, AI325039, AI663996, DT1P1A7, S100F
Chromosome Location:
Chr.3: 90537257 - 90543151 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01216812_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: S100a16
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_026416.2 NP_080692.1 1 - 2 95 99
GenBank mRNA BC020031.1 - 1 - 2 95 3980274 99

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the BC020031.1, NM_026416.2 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene S100a16

Panther Classification:

Molecular Function -

calmodulin-related

Gene Ontology Categories:

Function(s) Process(es)

response to calcium ion
calcium ion binding
protein homodimerization activity
poly(A) RNA binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0300006267.mm.1 NM_026416.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300006267.mm.1 NM_026416.2 Refseq
Affymetrix Mouse Clariom S Array PSR0300006267.mm.1 TC0300000802.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300006269.mm.1 NM_026416.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300006269.mm.1 NM_026416.2 Refseq
Affymetrix Mouse Clariom S Array PSR0300006269.mm.1 TC0300000802.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 917377 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0