This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01216279_g1
See other SLC4A1AP GE Assays ›
Gene Symbol:
Slc4a1ap
Gene Name:
solute carrier family 4 (anion exchanger), member 1, adaptor protein
Gene Aliases:
kanadaptin
Chromosome Location:
Chr.5: 31526850 - 31554038 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01216279_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slc4a1ap
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_009206.2 NP_033232.2 4 - 5 1069 72
GenBank mRNA AF035526.1 - 5 - 6 952 72
AK138398.1 - 4 - 5 1024 72
AK160250.1 - 3 - 4 944 72
BC072588.1 - 4 - 5 1052 72
BC090964.1 - 4 - 5 621 30625511 72

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500000325.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Slc4a1ap

Panther Classification:

Molecular Function -

RNA splicing factor

Gene Ontology Categories:

Function(s) Process(es)

biological_process
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500002909.mm.1 NM_009206.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500002909.mm.1 NM_009206.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500002909.mm.1 TC0500000325.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500002907.mm.1 NM_009206.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500002907.mm.1 NM_009206.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500002907.mm.1 TC0500000325.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0