Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0500022403.mm.1 |
NM_001201413.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500022403.mm.1 |
NM_001201413.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500022403.mm.1 |
NM_001201414.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500022403.mm.1 |
NM_001201414.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500022403.mm.1 |
NM_001201415.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500022403.mm.1 |
NM_001201415.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500022403.mm.1 |
NM_009686.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500022403.mm.1 |
TC0500002508.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500022404.mm.1 |
NM_001201413.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500022404.mm.1 |
NM_001201413.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500022404.mm.1 |
NM_009686.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500022404.mm.1 |
TC0500002508.mm.1 |
MTA TC |