This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01215808_m1
See other RIPK2 GE Assays ›
Gene Symbol:
Ripk2
Gene Name:
receptor (TNFRSF)-interacting serine-threonine kinase 2
Gene Aliases:
2210420D18Rik, CARD3, CARDIAK, CCK, D4Bwg0615e, RICK, RIP2
Chromosome Location:
Chr.4: 16122741 - 16163679 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01215808_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ripk2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_138952.3 NP_620402.1 8 - 9 1126 126
GenBank mRNA AF461040.1 - 8 - 9 1024 126
AF487539.1 - 8 - 9 1024 126
AK159593.1 - 8 - 9 1150 126
AK172319.1 - 8 - 9 1148 126
AK172417.1 - 8 - 9 1148 126
BC025611.1 - 6 - 7 644 126
BC050845.1 - 8 - 9 1140 126
BC069878.1 - 8 - 9 1151 5316066 126

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0400002251.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ripk2

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of cytokine-mediated signaling pathway
adaptive immune response
immune system process
positive regulation of T-helper 1 type immune response
protein phosphorylation
apoptotic process
I-kappaB kinase/NF-kappaB signaling
positive regulation of peptidyl-threonine phosphorylation
phosphorylation
positive regulation of protein ubiquitination
lipopolysaccharide-mediated signaling pathway
positive regulation of chemokine production
positive regulation of interferon-gamma production
positive regulation of interleukin-2 production
positive regulation of interleukin-6 production
positive regulation of tumor necrosis factor production
positive regulation of stress-activated MAPK cascade
positive regulation of immature T cell proliferation
positive regulation of peptidyl-serine phosphorylation
toll-like receptor 2 signaling pathway
toll-like receptor 4 signaling pathway
T cell proliferation
regulation of apoptotic process
positive regulation of apoptotic process
positive regulation of I-kappaB kinase/NF-kappaB signaling
regulation of cysteine-type endopeptidase activity involved in apoptotic process
response to exogenous dsRNA
innate immune response
positive regulation of T-helper 1 cell differentiation
positive regulation of JNK cascade
positive regulation of alpha-beta T cell proliferation
positive regulation of peptidyl-tyrosine phosphorylation
defense response to Gram-positive bacterium
T cell receptor signaling pathway
positive regulation of NF-kappaB transcription factor activity
positive regulation of ERK1 and ERK2 cascade
nucleotide-binding oligomerization domain containing 1 signaling pathway
nucleotide-binding oligomerization domain containing 2 signaling pathway
response to interleukin-1
response to interleukin-12
response to interleukin-18
cellular response to lipopolysaccharide
cellular response to lipoteichoic acid
cellular response to peptidoglycan
cellular response to muramyl dipeptide
nucleotide binding
protein kinase activity
protein serine/threonine kinase activity
non-membrane spanning protein tyrosine kinase activity
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
LIM domain binding
protein homodimerization activity
CARD domain binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0400018754.mm.1 NM_138952.4 Refseq
Affymetrix Mouse Clariom S Array PSR0400018754.mm.1 TC0400002251.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0