This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01215748_m1
See other CABP2 GE Assays ›
Gene Symbol:
Cabp2
Gene Name:
calcium binding protein 2
Gene Aliases:
-
Chromosome Location:
Chr.19: 4081469 - 4087340 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01215748_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cabp2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001160252.1 NP_001153724.1 1 - 2 98 98
GenBank mRNA AF169156.1 - 1 - 2 126 98

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AF169156.1, NM_001160252.1 transcripts.

Set Membership:

Amplicon length between 86 and 100 Mus musculus Probe spans exons

Gene Ontology Categories:

Process(es)

calcium ion binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1900000306.mm.1 NM_001160252.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900000306.mm.1 NM_001160252.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900000306.mm.1 NM_013878.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900000306.mm.1 NM_013878.2 Refseq
Affymetrix Mouse Clariom S Array PSR1900000306.mm.1 TC1900000025.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1900000308.mm.1 NM_001160252.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900000308.mm.1 NM_001160252.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900000308.mm.1 NM_001160253.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900000308.mm.1 NM_001160253.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900000308.mm.1 NM_013878.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900000308.mm.1 NM_013878.2 Refseq
Affymetrix Mouse Clariom S Array PSR1900000308.mm.1 TC1900000025.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0