This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01215743_m1
See other CABP2 GE Assays ›
Gene Symbol:
Cabp2
Gene Name:
calcium binding protein 2
Gene Aliases:
-
Chromosome Location:
Chr.19: 4081469 - 4087340 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01215743_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cabp2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001160253.1 NP_001153725.1 1 - 2 381 84
GenBank mRNA AK007488.1 - 1 - 2 382 84

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1900000025.mm.1

Set Membership:

Amplicon length between 71 and 85 Mus musculus Probe spans exons

Gene Ontology Categories:

Process(es)

calcium ion binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1900000303.mm.1 NM_001160253.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900000303.mm.1 NM_001160253.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900000303.mm.1 TC1900000025.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1900000304.mm.1 NM_001160253.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900000304.mm.1 NM_001160253.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900000304.mm.1 TC1900000025.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0