This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01215548_m1
See other TTC3 GE Assays ›
Gene Symbol:
Ttc3
Gene Name:
tetratricopeptide repeat domain 3
Gene Aliases:
2610202A04Rik, AA409221, D16Ium21, D16Ium21e, TPRD, mKIAA4119
Chromosome Location:
Chr.16: 94370739 - 94469222 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01215548_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ttc3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_009441.2 NP_033467.2 34 - 35 4551 65
GenBank mRNA AB008516.1 - 33 - 34 4428 65
AK148494.1 - 7 - 8 1575 65
AK161168.1 - 2 - 3 3464 65
AK164106.1 - 2 - 3 152 65
AK164492.1 - 2 - 3 1073 65
BC145489.1 - 29 - 30 4071 65

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1600001057.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ttc3

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Gene Ontology Categories:

Function(s) Process(es)

ubiquitin-dependent protein catabolic process
negative regulation of cell morphogenesis involved in differentiation
negative regulation of neuron differentiation
protein K48-linked ubiquitination
ubiquitin-protein transferase activity
protein binding
zinc ion binding
UDP-N-acetylmuramoylalanyl-D-glutamyl-2,6-diaminopimelate-D-alanyl-D-alanine ligase activity
ligase activity
ribosomal S6-glutamic acid ligase activity
coenzyme F420-0 gamma-glutamyl ligase activity
coenzyme F420-2 alpha-glutamyl ligase activity
metal ion binding
protein-glycine ligase activity
protein-glycine ligase activity, initiating
protein-glycine ligase activity, elongating
tubulin-glycine ligase activity
protein-glutamic acid ligase activity
tubulin-glutamic acid ligase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1600008349.mm.1 NM_009441.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600008349.mm.1 NM_009441.2 Refseq
Affymetrix Mouse Clariom S Array PSR1600008349.mm.1 TC1600001057.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1600008344.mm.1 NM_009441.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600008344.mm.1 NM_009441.2 Refseq
Affymetrix Mouse Clariom S Array PSR1600008344.mm.1 TC1600001057.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0