This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01215237_m1
See other TPD52L2 GE Assays ›
Gene Symbol:
Tpd52l2
Gene Name:
tumor protein D52-like 2
Gene Aliases:
2810411G23Rik, AU016537, C86069, D54
Chromosome Location:
Chr.2: 181497104 - 181517962 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01215237_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tpd52l2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001291197.1 NP_001278126.1 3 - 4 488 74
NM_001291200.1 NP_001278129.1 3 - 4 488 74
NM_025482.3 NP_079758.2 3 - 4 488 74
NR_111899.1 2 - 3 342 74
GenBank mRNA AK013085.1 - 3 - 4 430 74
AK013188.1 - 3 - 4 441 74
AK147885.1 - 3 - 4 448 74
AK159925.1 - 3 - 4 416 74
AK171825.1 - 3 - 4 450 74
BC006886.1 - 3 - 4 377 2647702 74
CN721218.1 - 2 - 3 342 74

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200002826.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tpd52l2

Gene Ontology Categories:

Function(s) Process(es)

biological_process
protein homodimerization activity
poly(A) RNA binding
protein heterodimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200023311.mm.1 NM_025482.3 Refseq
Affymetrix Mouse Clariom S Array PSR0200023311.mm.1 TC0200002826.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0