This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01215195_m1
See other UCKL1 GE Assays ›
Gene Symbol:
Uckl1
Gene Name:
uridine-cytidine kinase 1-like 1
Gene Aliases:
1110007H10Rik, Urkl1
Chromosome Location:
Chr.2: 181569149 - 181581973 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01215195_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Uckl1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_026765.3 NP_081041.2 8 - 9 928 102
GenBank mRNA BC016535.1 - 8 - 9 928 2655873 102

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Uckl1

Panther Classification:

Molecular Function -

nucleotide kinase

Gene Ontology Categories:

Function(s) Process(es)

biological_process
metabolic process
phosphorylation
nucleotide binding
uridine kinase activity
ATP binding
phosphofructokinase activity
phosphorylase kinase regulator activity
cobinamide kinase activity
phytol kinase activity
kinase activity
phosphatidylinositol phosphate kinase activity
cyclin-dependent protein serine/threonine kinase regulator activity
transferase activity
phenol kinase activity
cyclin-dependent protein kinase activating kinase regulator activity
inositol tetrakisphosphate 2-kinase activity
heptose 7-phosphate kinase activity
aminoglycoside phosphotransferase activity
phosphatidylinositol 3-kinase activity
eukaryotic elongation factor-2 kinase regulator activity
eukaryotic elongation factor-2 kinase activator activity
LPPG:FO 2-phospho-L-lactate transferase activity
cytidine kinase activity
glycerate 2-kinase activity
(S)-lactate 2-kinase activity
phosphoserine:homoserine phosphotransferase activity
L-seryl-tRNA(Sec) kinase activity
phosphocholine transferase activity
polynucleotide 5'-hydroxyl-kinase activity
ATP-dependent polynucleotide kinase activity
GTP-dependent polynucleotide kinase activity
farnesol kinase activity
CTP:2-trans,-6-trans-farnesol kinase activity
geraniol kinase activity
geranylgeraniol kinase activity
CTP:geranylgeraniol kinase activity
prenol kinase activity
1-phosphatidylinositol-5-kinase activity
1-phosphatidylinositol-3-phosphate 4-kinase activity
phosphatidylinositol-3,4-bisphosphate 5-kinase activity
phosphatidylinositol bisphosphate kinase activity
inositol-3,4,6-trisphosphate 1-kinase activity
inositol 5-diphosphate pentakisphosphate 5-kinase activity
inositol diphosphate tetrakisphosphate kinase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0