This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01215026_g1
See other HELZ2 GE Assays ›
Gene Symbol:
Helz2
Gene Name:
helicase with zinc finger 2, transcriptional coactivator
Gene Aliases:
PDIP1, Pric285, mKIAA1769, mPDIP1
Chromosome Location:
Chr.2: 181227615 - 181242058 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01215026_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Helz2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_183162.2 NP_898985.2 13 - 14 7868 60
GenBank mRNA AB201714.1 - 12 - 13 7772 60
AK173259.1 - 2 - 3 5150 60
BC006779.1 - 5 - 6 976 60

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200005438.mm.1

Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

DNA metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
nucleotide binding
DNA binding
helicase activity
ATP binding
hydrolase activity
ligand-dependent nuclear receptor transcription coactivator activity
poly(A) RNA binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200046352.mm.1 NM_183162.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200046352.mm.1 NM_183162.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200046352.mm.1 TC0200005438.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0