This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01214279_m1
See other C330002G04RIK GE Assays ›
Gene Symbol:
C330002G04Rik
Gene Name:
RIKEN cDNA C330002G04 gene
Gene Aliases:
-
Chromosome Location:
Chr.19: 22996890 - 23075906 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01214279_m1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: C330002G04Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA BC044868.1 - 4 - 5 405 68

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream transient receptor potential cation channel,
subfamily M,
member 3
Trpm3 6330504P12Rik,
9330180E14,
AU018608,
B930001P07Rik,
LTRPC3,
MLSN2
-71.9kb Anti-sense Chr. 19 - 22138181 - 22989897 on NCBI Build GRCm38
Downstream Kruppel-like factor 9 Klf9 2310051E17Rik,
AA589643,
BTEB-1,
Bteb1,
ENSMUSG00000055440,
Gm9971
77.8kb Anti-sense Chr. 19 - 23141226 - 23166911 on NCBI Build GRCm38

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons

MicroArray Probes:

Agilent ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 598391 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 362049 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 538159 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 557431 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 60605 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 148641 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 611438 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 813247 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 592724 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 17368 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 29646 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 746474 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 86433 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 732960 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 658157 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 540749 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 446802 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 114967 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 897856 NM_001111282.1 refseq_mrna_mouse_NM
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 897856 XM_485920.4 refseq_mrna_mouse_XM
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 897856 XM_916659.3 refseq_mrna_mouse_XM
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 348439 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 409845 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 129765 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 222148 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 529196 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 272273 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 728923 AK012067.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 728923 AK029107.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 728923 AK082303.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 728923 BC003981.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 728923 NM_028120.2 refseq_mrna_mouse_NM
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 728923 XM_911662.2 refseq_mrna_mouse_XM
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 869432 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 140281 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 43882 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 769214 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 72332 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 801737 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 945413 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 584101 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 703334 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 712208 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 84178 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 211007 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 495232 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 13485 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 123618 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 64537 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 114216 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 22530 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P132813 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09131341 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03567979 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P132813 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09131209 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09131241 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09131226 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09131304 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P09131341 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P132813 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P03567979 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14296223 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14364253 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015472 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015659 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015641 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015745 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015502 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015707 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015725 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015650 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015686 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015627 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015688 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14173442 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015695 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14501888 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015596 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015642 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015684 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015690 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015762 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015654 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015633 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015705 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14173438 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015687 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14176771 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015699 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015724 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015771 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015480 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015774 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015618 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015759 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015537 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14141297 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015747 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015477 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015608 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14413793 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14164673 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015648 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14159234 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015763 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14367790 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015513 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14319197 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015760 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015446 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015719 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015752 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14276860 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015544 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015693 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015628 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015677 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015743 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015673 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015746 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14463705 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015399 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015753 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015500 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015644 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015779 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015619 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015755 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015672 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015656 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14423371 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14503991 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015731 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015540 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015710 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015505 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14203721 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14364249 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015542 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015456 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015657 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015679 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015714 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015727 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015602 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015449 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015541 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015756 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015459 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015393 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015670 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14459211 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015450 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015481 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015647 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015702 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015600 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015691 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015715 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14276858 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015511 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14141301 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14372437 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015768 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015669 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015783 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015708 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015767 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14214825 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015443 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14225258 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015509 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015454 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14197413 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015534 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015630 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015786 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015664 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015736 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015452 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015598 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015728 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015704 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015744 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015631 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14229410 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015605 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14413798 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015503 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14441305 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015740 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14497382 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015507 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015404 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015717 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015722 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015666 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015501 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015451 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015778 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015594 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14276863 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015703 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015397 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015394 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015506 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015711 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015652 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14267372 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32015751 - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 534860 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 103170452 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0