This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01214223_g1
See other GNL3 GE Assays ›
Gene Symbol:
Gnl3
Gene Name:
guanine nucleotide binding protein-like 3 (nucleolar)
Gene Aliases:
Ns
Chromosome Location:
Chr.14: 31012433 - 31019131 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01214223_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gnl3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_153547.6 NP_705775.2 9 - 10 1039 93
NR_104298.1 9 - 10 991 93
GenBank mRNA AK077523.1 - 9 - 10 970 93
AK167183.1 - 9 - 10 923 93
AY181025.1 - 9 - 10 897 93
AY185498.1 - 9 - 10 897 93
BC037996.1 - 9 - 10 899 5052858 93

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1400001665.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gnl3

Gene Ontology Categories:

Function(s) Process(es)

cell proliferation
regulation of cell proliferation
ribosome biogenesis
positive regulation of pri-miRNA transcription from RNA polymerase II promoter
nucleotide binding
GTPase activity
protein binding
GTP binding
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1400012637.mm.1 NM_153547.6 Refseq
Affymetrix Mouse Clariom S Array PSR1400012637.mm.1 NR_104298.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400012637.mm.1 TC1400001665.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0