This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01214209_m1
See other NEK4 GE Assays ›
Gene Symbol:
Nek4
Gene Name:
NIMA (never in mitosis gene a)-related expressed kinase 4
Gene Aliases:
Stk2
Chromosome Location:
Chr.14: 30951382 - 30988476 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01214209_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Nek4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AF099067.1 - 2 - 3 492 125
AJ223071.1 - 2 - 3 582 125
AK078809.1 - 2 - 3 604 125
AK132803.1 - 2 - 3 520 125
AK144130.1 - 2 - 3 466 125
AK147381.1 - 2 - 3 570 125
AK149584.1 - 2 - 3 520 125
AK168094.1 - 2 - 3 503 125
BC057939.1 - 2 - 3 494 5325558 125
Y09234.1 - 2 - 3 582 125

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-655dcf6cc8-78hlp:80/100.66.128.142:80.
git-commit: 0d764ad91c70359c551b4d51a80efc6847a0ee54
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0