This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01213859_m1
See other RFC3 GE Assays ›
Gene Symbol:
Rfc3
Gene Name:
replication factor C (activator 1) 3
Gene Aliases:
2810416I22Rik, 38kDa, AU022547, Recc3
Chromosome Location:
Chr.5: 151642824 - 151651208 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01213859_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rfc3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_027009.2 NP_081285.1 1 - 2 144 97
GenBank mRNA AK013095.1 - 1 - 2 97
AK042393.1 - 1 - 2 145 97
AK145126.1 - 1 - 2 171 97
AK165891.1 - 1 - 2 139 97
AK167047.1 - 1 - 2 145 97
BC026795.1 - 1 - 2 115 4015134 97

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500003705.mm.1

Set Membership:

Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Panther Classification:

Molecular Function -

DNA-directed DNA polymerase

Gene Ontology Categories:

Function(s) Process(es)

DNA replication
DNA-dependent DNA replication
DNA binding
DNA clamp loader activity
ATPase activity

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500033805.mm.1 NM_027009.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500033805.mm.1 NM_027009.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500033805.mm.1 TC0500003705.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500033806.mm.1 NM_027009.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500033806.mm.1 NM_027009.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500033806.mm.1 TC0500003705.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 760949 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0